ENA432539367 Enamine

2-(dimethyl-1,2-oxazol-4-yl)acetonitrile

  • MDL number MFCD08701345
  • Cas Number : 35166-42-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195627

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195627
Availability details for
Quantity
  • Availability
  • Add to Cart