ENA432539376 Enamine

N'-(pyrrolidin-3-yl)-2-(4H-1,2,4-triazol-3-yl)ethanimidamide trihydrochloride

  • Cas Number : 1807938-75-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 3HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195639

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195639
Availability details for
Quantity
  • Availability
  • Add to Cart