ENA432539380 Enamine

1-(2,6-dichlorophenyl)-1H-1,2,3-triazole-5-carboxylic acid

  • MDL number MFCD21242752
  • Cas Number : 1492720-78-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195645

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195645
Availability details for
Quantity
  • Availability
  • Add to Cart