ENA432539381 Enamine

4-(dimethylamino)-2-methylpyrimidine-5-carbaldehyde

  • MDL number MFCD28145276
  • Cas Number : 1545329-39-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195646

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195646
Availability details for
Quantity
  • Availability
  • Add to Cart