ENA432539388 Enamine

4-(4-chlorophenyl)piperazine-1-carboxamide

  • MDL number MFCD04153414
  • Cas Number : 676579-72-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195011

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195011
Availability details for
Quantity
  • Availability
  • Add to Cart