ENA432539391 Enamine

N2-[2-(4-fluorophenyl)ethyl]-1,3-benzoxazole-2,6-diamine

  • MDL number MFCD28139641
  • Cas Number : 1519179-15-7
  • Molecular Weight : 271.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N2-(4-fluorophenethyl)benzo[d]oxazole-2,6-diamine
Inchi Code 1S/C15H14FN3O/c16-11-3-1-10(2-4-11)7-8-18-15-19-13-6-5-12(17)9-14(13)20-15/h1-6,9H,7-8,17H2,(H,18,19)
Inchi Key WPUBWYHCIXWBHQ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-195673

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195673
Availability details for
Quantity
  • Availability
  • Add to Cart