ENA432539395 Enamine

{[1-(3-bromophenyl)-1H-1,2,3-triazol-4-yl]methyl}(methyl)amine hydrochloride

  • MDL number MFCD28139643
  • Cas Number : 1803588-96-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-195679

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195679
Availability details for
Quantity
  • Availability
  • Add to Cart