ENA432539397 Enamine

1-phenylethane-1,2-diamine dihydrochloride

  • MDL number MFCD01726798
  • Cas Number : 16635-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-195683

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195683
Availability details for
Quantity
  • Availability
  • Add to Cart