ENA432539403 Enamine

4-chloro-2-hydroxybenzene-1-sulfonamide

  • MDL number MFCD16999674
  • Cas Number : 1243372-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-195740

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195740
Availability details for
Quantity
  • Availability
  • Add to Cart