ENA432539405 Enamine

(2R)-2-aminobutane-1,4-diol

  • MDL number MFCD18205987
  • Cas Number : 80572-06-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195743

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195743
Availability details for
Quantity
  • Availability
  • Add to Cart