ENA432539407 Enamine

methyl 2-(prop-2-enamido)benzoate

  • MDL number MFCD12091604
  • Cas Number : 104774-81-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-195745

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195745
Availability details for
Quantity
  • Availability
  • Add to Cart