ENA432539412 Enamine

N-cycloheptyl-2-(4,5-dihydro-1H-imidazol-2-ylsulfanyl)acetamide hydrochloride

  • MDL number MFCD28145281
  • Cas Number : 1803605-68-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195751

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195751
Availability details for
Quantity
  • Availability
  • Add to Cart