ENA432539417 Enamine

5-(bromomethyl)pyrrolidin-2-one

  • MDL number MFCD04038517
  • Cas Number : 190271-85-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195756

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195756
Availability details for
Quantity
  • Availability
  • Add to Cart