ENA432539418 Enamine

5-ethynylpyridine-3-carboxamide

  • MDL number MFCD18254891
  • Cas Number : 1256818-70-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195757

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195757
Availability details for
Quantity
  • Availability
  • Add to Cart