ENA432539421 Enamine

1-(3,4-dichlorophenyl)-1H-imidazole-4-carboxylic acid hydrochloride

  • MDL number MFCD28145282
  • Cas Number : 1803601-01-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-195761
Availability details for
Quantity
  • Availability
  • Add to Cart