ENA432539422 Enamine

2-amino-2-(5-bromo-2-fluorophenyl)propanoic acid

  • MDL number MFCD18696039
  • Cas Number : 1266784-23-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-195764

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195764
Availability details for
Quantity
  • Availability
  • Add to Cart