ENA432539425 Enamine

3-fluoro-4-(1,3,4-oxadiazol-2-yl)aniline

  • MDL number MFCD16084693
  • Cas Number : 1249768-66-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-195767

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195767
Availability details for
Quantity
  • Availability
  • Add to Cart