ENA432539426 Enamine

methyl (2S)-2-[(furan-2-yl)formamido]propanoate

  • MDL number MFCD20358342
  • Cas Number : 17107-27-6
  • Molecular Weight : 197.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl (2S)-2-(2-furoylamino)propanoate
Inchi Code 1S/C9H11NO4/c1-6(9(12)13-2)10-8(11)7-4-3-5-14-7/h3-6H,1-2H3,(H,10,11)/t6-/m0/s1
Inchi Key WNQAJUYLFLETEV-LURJTMIESA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-195779

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195779
Availability details for
Quantity
  • Availability
  • Add to Cart