ENA432539427 Enamine

3-(4-amino-3-methylphenoxy)-1-methylpyrrolidin-2-one

  • MDL number MFCD20284701
  • Cas Number : 1343823-13-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195786

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195786
Availability details for
Quantity
  • Availability
  • Add to Cart