ENA432539431 Enamine

2-amino-5,5-dimethylhexan-1-ol hydrochloride

  • MDL number MFCD08455585
  • Cas Number : 1314930-59-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195026

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195026
Availability details for
Quantity
  • Availability
  • Add to Cart