ENA432539435 Enamine

2-(difluoromethyl)-1-methyl-1H-1,3-benzodiazole-5-carboxylic acid

  • MDL number MFCD19601492
  • Cas Number : 1339147-00-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195838

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195838
Availability details for
Quantity
  • Availability
  • Add to Cart