ENA432539438 Enamine

4-bromocyclohexane-1-carboxamide

  • MDL number MFCD16877239
  • Cas Number : 1360438-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195847

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195847
Availability details for
Quantity
  • Availability
  • Add to Cart