ENA432539446 Enamine

5-bromo-7-methoxy-2-methyl-3,4-dihydroquinazolin-4-one

  • MDL number MFCD28145288
  • Cas Number : 1593360-88-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-195873

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195873
Availability details for
Quantity
  • Availability
  • Add to Cart