ENA432539460 Enamine

methyl 4-(propan-2-yloxy)cyclohexane-1-carboxylate

  • MDL number MFCD23134171
  • Cas Number : 1224582-23-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-195891

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195891
Availability details for
Quantity
  • Availability
  • Add to Cart