ENA432539461 Enamine

2,2,4-trimethyl-8-(propan-2-yl)-1,2,3,4-tetrahydroquinoline

  • MDL number MFCD24618912
  • Cas Number : 1333235-93-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195894

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195894
Availability details for
Quantity
  • Availability
  • Add to Cart