ENA432539465 Enamine

6-chloro-4-iodo-3-methoxypyridazine

  • MDL number MFCD12827448
  • Cas Number : 181355-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195913

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195913
Availability details for
Quantity
  • Availability
  • Add to Cart