ENA432539470 Enamine

imidazo[1,2-a]pyridine-8-carbaldehyde

  • MDL number MFCD06739248
  • Cas Number : 136117-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195925

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195925
Availability details for
Quantity
  • Availability
  • Add to Cart