ENA432539475 Enamine

2-methyl-5-(morpholin-4-yl)aniline

  • MDL number MFCD11935300
  • Cas Number : 1007211-91-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195934

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195934
Availability details for
Quantity
  • Availability
  • Add to Cart