ENA432539482 Enamine

4-chloro-N,N-dimethyl-1,2,5-thiadiazol-3-amine

  • MDL number MFCD09036497
  • Cas Number : 937675-08-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194897

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194897
Availability details for
Quantity
  • Availability
  • Add to Cart