ENA432539486 Enamine

5-chloro-2-methyl-1,3-benzothiazol-6-amine

  • MDL number MFCD18826625
  • Cas Number : 101253-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-195050

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195050
Availability details for
Quantity
  • Availability
  • Add to Cart