ENA432539489 Enamine

3-(3-amino-4-methylphenyl)-4,5-dihydro-1H-1,2,4-triazol-5-one

  • MDL number MFCD14640875
  • Cas Number : 1248756-57-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195976

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195976
Availability details for
Quantity
  • Availability
  • Add to Cart