ENA432539492 Enamine

6-[methyl(phenyl)amino]pyridine-3-carbonitrile

  • MDL number MFCD09938161
  • Cas Number : 1016885-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-195064
Availability details for
Quantity
  • Availability
  • Add to Cart