ENA432539497 Enamine

trans-1-tert-butyl-2-[1-(2-methoxyethyl)-1H-pyrazol-4-yl]-5-oxopyrrolidine-3-carbonitrile

  • MDL number MFCD28657086
  • Cas Number : 1808696-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-196010
Availability details for
Quantity
  • Availability
  • Add to Cart