ENA432539498 Enamine

3-carbamoyl-2-(3-methylbutanamido)propanoic acid

  • MDL number MFCD09050681
  • Cas Number : 880479-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196011

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196011
Availability details for
Quantity
  • Availability
  • Add to Cart