ENA432539499 Enamine

N-methyl-N-phenylpyrrolidine-3-carboxamide hydrochloride

  • MDL number MFCD28145304
  • Cas Number : 1579171-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196013

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196013
Availability details for
Quantity
  • Availability
  • Add to Cart