ENA432539505 Enamine

5-ethyl-2-phenyl-1H-imidazole-4-carboxylic acid

  • MDL number MFCD12171772
  • Cas Number : 1156729-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196019

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196019
Availability details for
Quantity
  • Availability
  • Add to Cart