ENA432539512 Enamine

5-(4-methoxyphenyl)pyrimidin-4-amine

  • MDL number MFCD07381992
  • Cas Number : 35202-24-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196026

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196026
Availability details for
Quantity
  • Availability
  • Add to Cart