ENA432539513 Enamine

5-(1-methyl-1H-imidazol-2-yl)-4-phenyl-1,3-thiazol-2-amine

  • MDL number MFCD09040421
  • Cas Number : 929975-34-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196027

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196027
Availability details for
Quantity
  • Availability
  • Add to Cart