ENA432539517 Enamine

6-chloro-2-methyl-[1,2,4]triazolo[1,5-b]pyridazine

  • MDL number MFCD11044767
  • Cas Number : 55643-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195071

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195071
Availability details for
Quantity
  • Availability
  • Add to Cart