ENA432539519 Enamine

N-{3-[(3-bromopyridin-2-yl)amino]phenyl}acetamide

  • MDL number MFCD13476704
  • Cas Number : 1293156-74-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196040

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196040
Availability details for
Quantity
  • Availability
  • Add to Cart