ENA432539522 Enamine

1-(aminomethyl)cyclopropane-1-carbonitrile hydrochloride

  • MDL number MFCD19382420
  • Cas Number : 1205544-78-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196047

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196047
Availability details for
Quantity
  • Availability
  • Add to Cart