ENA432539527 Enamine

cyclooctylmethanamine

  • MDL number MFCD00154120
  • Cas Number : 4734-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196054

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196054
Availability details for
Quantity
  • Availability
  • Add to Cart