ENA432539529 Enamine

iminodimethyl-lambda6-sulfanone

  • MDL number MFCD02656633
  • Cas Number : 1520-31-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 51-58
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196070

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P312,P330,P363,P501
hcodes Xn
MSDS https://www.enaminestore.com/catalog/EN300-196070
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart