ENA432539530 Enamine

2-methyl-3-phenylcyclopropane-1-carboxylic acid

  • MDL number MFCD20643770
  • Cas Number : 24581-87-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196073

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196073
Availability details for
Quantity
  • Availability
  • Add to Cart