ENA432539532 Enamine

5-bromo-2-chloro-N-(propan-2-yl)pyridin-3-amine

  • MDL number MFCD22628164
  • Cas Number : 1112982-73-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196082

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196082
Availability details for
Quantity
  • Availability
  • Add to Cart