ENA432539534 Enamine

1-[2-hydroxy-3-(trifluoromethyl)phenyl]ethan-1-one

  • MDL number MFCD18397605
  • Cas Number : 1144501-67-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196085

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196085
Availability details for
Quantity
  • Availability
  • Add to Cart