ENA432539536 Enamine

2-(1-ethyl-1H-imidazol-2-yl)piperidine

  • MDL number MFCD19622462
  • Cas Number : 1344365-73-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-196088

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196088
Availability details for
Quantity
  • Availability
  • Add to Cart