ENA432539537 Enamine

N-methyl-N-(2-methylpropyl)carbamoyl chloride

  • MDL number MFCD23790134
  • Cas Number : 1174330-61-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-196089

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196089
Availability details for
Quantity
  • Availability
  • Add to Cart