ENA432539540 Enamine

[(3-fluorophenyl)amino]formonitrile

  • MDL number MFCD16767978
  • Cas Number : 14213-18-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196092

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196092
Availability details for
Quantity
  • Availability
  • Add to Cart