ENA432539541 Enamine

[(2-fluorophenyl)amino]formonitrile

  • MDL number MFCD12402222
  • Cas Number : 71232-23-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196093

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196093
Availability details for
Quantity
  • Availability
  • Add to Cart